Anti-E2F1 Mouse Monoclonal Antibody [clone: KH95]

Supplier: GeneTex
GTX24070
89352-106EA 677.42 USD
89352-106
Anti-E2F1 Mouse Monoclonal Antibody [clone: KH95]
Antibodies
Mouse monoclonal [KH95] to E2F1

Immunohistochemistry: use at a dilution of 1/25 - 1/50 for 30mins at RT (ABC method). Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. For formalin fixed paraffin embedded sections, perform heat mediated antigen retrieval using 10mM citrate buffer pH 6.0 before commencing with the IHC staining protocol.

Type: Primary
Antigen: E2F1
Clonality: Monoclonal
Clone: KH95
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR