Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CK8 Mouse Monoclonal Antibody [clone: 35bE11]

Supplier: Genetex

GTX27506
89362-596EA 325.11 USD InStock
89362-596
Anti-CK8 Mouse Monoclonal Antibody [clone: 35bE11]
Antibodies
Mouse monoclonal [35bE11] to Cytokeratin 8 prediluted

IHC-P: Use at an assay dependent dilution. IHC-Fr: Use at an assay dependent dilution. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions alland the staining system employed, optimal incubation should be determined by the user. Enzymatic predigestion of paraffin embedded tissues is recommended prior to the immunostaining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: 35bE11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty