Anti-CD82 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100633
89348-582EA 557.66 USD
89348-582
Anti-CD82 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD82 (CD82 molecule)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD82
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR