Anti-CD8 Rat Monoclonal Antibody [clone: YTC141.1HL]

GTX76097
89367-958EA 376.99 USD
89367-958
Anti-CD8 Rat Monoclonal Antibody [clone: YTC141.1HL]
Rat monoclonal antibody [YTC141.1HL] to CD8

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR