Anti-CD8 Mouse Monoclonal Antibody [clone: C8/144B]

Supplier: GeneTex
GTX72053
89352-940EA 679.12 USD
89352-940
Anti-CD8 Mouse Monoclonal Antibody [clone: C8/144B]
Antibodies
Mouse Monoclonal to CD8 T - cell * For Paraffin Sections

This antibody can be used on formalin-fixed, paraffin-embedded tissue sections, as well as on frozen tissue sections. For a three-step streptavidin-biotin complex detection system a dilution of 1:25 is recommended as a guideline. When used in conjunction with our LAB/Probe kits. To enhance staining , high temperature-based antigenic unmasking technique (see instructions below) is highly recommended when using paraffin-embedded, formalin-fixed sections. The optimal conditions should be determined by the individual laboratory.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR