Anti-TFR1 Mouse Monoclonal Antibody [clone: MEM-75]

Supplier: GeneTex
GTX78312
89353-830EA 457.04 USD
89353-830
Anti-TFR1 Mouse Monoclonal Antibody [clone: MEM-75]
Antibodies
Monoclonal Antibody to CD71 (Human) Fluorescein (FITC)

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 10^6 cells in a suspension. The content of a vial (2 ml) is sufficient for 100 tests.

Type: Primary
Antigen: TFR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR