Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD62E Mouse Monoclonal Antibody [clone: CL2/6]

Supplier: Genetex

GTX75665
89365-866EA 434.62 USD InStock
89365-866
Anti-CD62E Mouse Monoclonal Antibody [clone: CL2/6]
Antibodies
Mouse monoclonal antibody [CL2/6] to CD62E

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 1/20 Immunohistology - Frozen: 0.5 ug/ml - 2.0 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD62E
Clonality: Monoclonal
Clone: CL2/6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty