Anti-EVX1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77912
89366-414EA 619.97 USD
89366-414
Anti-EVX1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to EVX1

Type: Primary
Antigen: EVX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR