Anti-CRHR1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX88961
89366-970EA 577.91 USD
89366-970
Anti-CRHR1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CRHR1 / CRF-R (aa 107 - 117)

For ELISA (Peptide): Use at a starting dilution of 1:32,000. For IF (Mouse only): Use at a concentration of 2µg/ml. For WB (Human only): Use at a starting concentration of 0.3-0.3 µg/ml. Approx 60-70kDa band observed in human brain (cerebellum) and human colon lysates (calculated MW of 47.7kDa according to NP_004373.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Xu et al, Mol Pharmacol. 2001 Mar;59(3):485-92; PMID: 11179443). Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CRHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 107 - 117 aa
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR