Anti-CD8 Mouse Monoclonal Antibody [clone: LT8]

Supplier: GeneTex
GTX74777
89353-326EA 628.11 USD
89353-326
Anti-CD8 Mouse Monoclonal Antibody [clone: LT8]
Antibodies
Mouse monoclonal antibody [LT8] to CD8

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR