Anti-ITGB3 Mouse Monoclonal Antibody [clone: PM6/13]

GTX76440
89366-100EA 497.96 USD
89366-100
Anti-ITGB3 Mouse Monoclonal Antibody [clone: PM6/13]
Mouse monoclonal antibody [PM6/13] to Integrin beta 3

Recommended Starting Dilutions: Flow Cytometry: 1/100 - 1/200 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR