Anti-ITGB3 Mouse Monoclonal Antibody

Supplier: GeneTex

GTX75342
89365-764EA 450.98 USD
89365-764
Anti-ITGB3 Mouse Monoclonal Antibody
Antibodies
Recognises the rat CD61 cell surface antigen, also known as the integrin b3 chain.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR