Mouse Monoclonal to CD57 HNK - 1
Immunofluorescence 5-20 ug/mL. Flow cytometry 1-5 ug/10^6 cells. Immunoblotting 1-5 ug/mL. This antibody can be used on frozen cryostat sections as well as formalin-fixed, paraffin-embedded tissue sections. Prolonged fixation is buffered formalin can destroy the epitope. The antibody may be used at a dilution of 5-10 ug/mL with the Auto/Probe III kit. For consistent and reproducible results on formalin-fixed, paraffin-embedded sections, a high temperature antigen unmasking technique is required (see instructions below).
Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: B321 (NK-1)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human