Anti-NCAM1 Mouse Monoclonal Antibody [clone: T199]

GTX72033
89352-932EA 376.78 USD
89352-932
Anti-NCAM1 Mouse Monoclonal Antibody [clone: T199]
Mouse Monoclonal to CD56 Natural Killer Cell

Immunohistochemistry (Frozen), Flow Cytometry. For a three-step avidin-biotin complex system, a dilution of 1:25 to 1:50 may be used as a guideline. The optimal conditions should be determined by the individual laboratory. Dilute according to the particular application being used. In general, the 0.05M Borate pH 8.0 containing 0.15M Sodium Chloride, 0.05% Sodium Azide, is a good diluent to use with most antibodies.

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: T199
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR