Anti-ICAM1 Mouse Monoclonal Antibody [clone: 1H4]

Supplier: Genetex

GTX78387
89366-618EA 296.25 USD
89366-618
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 1H4]
Antibodies
Mouse monoclonal [1H4] to CD54 (Azide Free)

IHC-P: Recommended dilution: 10-20 ug/ml Application Note: Prolonged fixation in buffered formalin can destroy the epitope. Hight temperature antigen unmasking technique is required. IHC-Fr: Recommended dilution: 5-20 ug/ml.

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR