Anti-CD52 Rat Monoclonal Antibody [clone: YTH34.5]

Supplier: GeneTex
GTX43049
89363-522EA 480.08 USD
89363-522
Anti-CD52 Rat Monoclonal Antibody [clone: YTH34.5]
Antibodies
Rat monoclonal antibody [YTH34.5] to CD52

Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR