Anti-CD51/61 Mouse Monoclonal Antibody [clone: 23C6]

Supplier: GeneTex
GTX76488
89366-120EA 579.18 USD
89366-120
Anti-CD51/61 Mouse Monoclonal Antibody [clone: 23C6]
Antibodies
Mouse monoclonal antibody [23C6] to Integrin alpha V + beta 3

Recommended Starting Dilutions: Flow Cytometry: 1/40 - 1/80 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD51/61
Clonality: Monoclonal
Clone: 23C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR