Anti-CD5 Rabbit Monoclonal Antibody [clone: SP19]

Supplier: GeneTex
GTX79414
89367-182EA 1154.33 USD
89367-182
Anti-CD5 Rabbit Monoclonal Antibody [clone: SP19]
Antibodies
Monoclonal Rabbit Antibody to Human CD5

We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR