Anti-ITGA3 Mouse Monoclonal Antibody [clone: 17C6]

Supplier: GeneTex
GTX75610
89365-842EA 520.52 USD
89365-842
Anti-ITGA3 Mouse Monoclonal Antibody [clone: 17C6]
Antibodies
Mouse monoclonal antibody [17C6] to Integrin alpha 3

Type: Primary
Antigen: ITGA3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR