Anti-ITGA2 Rat Monoclonal Antibody [clone: DX5]

Supplier: GeneTex
GTX75859
89353-376EA 617.49 USD
89353-376
Anti-ITGA2 Rat Monoclonal Antibody [clone: DX5]
Antibodies
Rat monoclonal antibody [DX5] to Integrin alpha 2

Type: Primary
Antigen: ITGA2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR