Anti-CD47 Mouse Monoclonal Antibody [clone: BRIC126]

Supplier: Genetex

GTX76615
89366-156EA 610.77 USD
89366-156
Anti-CD47 Mouse Monoclonal Antibody [clone: BRIC126]
Antibodies
Mouse monoclonal antibody [BRIC126] to CD47

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: BRIC126
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR