Anti-PTPRC Mouse Monoclonal Antibody [clone: MEM-28]

Supplier: GeneTex
GTX78292
89366-584EA 606.79 USD
89366-584
Anti-PTPRC Mouse Monoclonal Antibody [clone: MEM-28]
Antibodies
Mouse monoclonal [MEM-28] to CD45 (Biotin)

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:500. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR