Anti-CD44V7 Mouse Monoclonal Antibody

GTX75294
89367-854EA 471.9 USD
89367-854
Anti-CD44V7 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody [VFF-9] to CD44v7

Type: Primary
Antigen: CD44V7
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR