Anti-CD44 Mouse Monoclonal Antibody [clone: OX-50]

Supplier: Genetex

GTX76382
89367-174EA 480.08 USD
89367-174
Anti-CD44 Mouse Monoclonal Antibody [clone: OX-50]
Antibodies
Mouse monoclonal antibody [OX-50] to CD44

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR