Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GP9 Mouse Monoclonal Antibody

Supplier: Genetex

GTX76338
89366-060EA 409.2 USD InStock
89366-060
Anti-GP9 Mouse Monoclonal Antibody
Antibodies
Mouse monoclonal antibody [FMC-25] to CD42a

Type: Primary
Antigen: GP9
Clonality: Monoclonal
Clone: FMC-25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty