Anti-CD41 Mouse Monoclonal Antibody [clone: PM6/248]

Supplier: GeneTex
GTX76180
89366-004EA 668.1 USD
89366-004
Anti-CD41 Mouse Monoclonal Antibody [clone: PM6/248]
Antibodies
Mouse monoclonal antibody [PM6/248] to CD41

Recommended Starting Dilutions: Flow Cytometry: 1/100 - 1/200 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR