Anti-CD41 Rat Monoclonal Antibody [clone: MWReg30]

Supplier: GeneTex
GTX76010
89365-956EA 772.72 USD
89365-956
Anti-CD41 Rat Monoclonal Antibody [clone: MWReg30]
Antibodies
Rat monoclonal antibody [MWReg30] to CD41

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR