Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-CD41 Mouse Monoclonal Antibody [clone: MEM-06]

Supplier: Genetex

GTX78346
89366-608EA 333.47 USD InStock
89366-608
Anti-CD41 Mouse Monoclonal Antibody [clone: MEM-06]
Antibodies
Monoclonal Antibody to CD41 (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry. Suggested working dilution is 1:200. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: MEM-06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty