CD40 ligand (CD40L), also known as CD154, gp39 and TRAP, is a 33 kDa type II membrane glycoprotein expressed mainly on the cell surface of activated T lymphocytes, but also exists as a soluble form extracellularly. CD40L is the ligand for CD40, a member of the TNF superfamily, which is expressed on the cell surface of B cells, macrophages/monocytes, dendritic cells, vascular endothelial cells, and epithelial cells. CD40L plays an important role in B cell proliferation, antibody class switching, modulation of apoptosis in the germinal center through interaction with B cells expressing CD40, and activation of CD4+ T cells. Mutation within the CD40L gene is linked to hyper IgM syndrome, an X-linked immunodeficiency disease that is characterized by elevated level of serum IgM and decreased level of other isotypes. CD40, also known as bp50 or CDW40, is a type I membrane protein found on the surface of B cells and primary carcinomas. It belongs to the TNF-R family. CD40 serves as the receptor for CD40 ligand (CD40L), a cytokine expressed on the cell surface of T cells. CD40 plays a critical role in B cell proliferation, antibody class switching, modulation of apoptosis in the germinal center through interaction with T cells expressing CD40L, and activation of CD4+ T cells. Ligation of CD40 with its ligand preferentially induces stress-activated protein kinases in B lymphocytes.
WB: Use at a concentration of 0.5 - 1 ?g/ml. ELISA: Use at a concentration of 2-3 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD40L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: