Anti-CD40 Rat Monoclonal Antibody [clone: 3/23]

Supplier: GeneTex
GTX74688
89365-534EA 241.03 USD
89365-534
Anti-CD40 Rat Monoclonal Antibody [clone: 3/23]
Antibodies
Rat monoclonal antibody [3/23] to CD40

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR