Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD4 Rat Monoclonal Antibody

Supplier: Genetex
GTX75337
89365-758
Anti-CD4 Rat Monoclonal Antibody
Rat monoclonal [YTS191.1] to CD4 (FITC)

For FACS: Use 1μg for 10^6 cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty