Anti-CD4 Rat Monoclonal Antibody [clone: YTS191.1]

Supplier: GeneTex
GTX75336
89365-756EA 592.61 USD
89365-756
Anti-CD4 Rat Monoclonal Antibody [clone: YTS191.1]
Antibodies
Rat monoclonal [YTS191.1] to mouse CD4 (Biotin)

For FACS: Use at a concentration of 0.1 ?g to label 10^6 cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR