Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]

Supplier: GeneTex
GTX74820
89365-590EA 608.82 USD
89365-590
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]
Antibodies
Mouse monoclonal [RPA-T4] to CD4 (azide free)

  • For FACS: Use at a dilution of 1:50 - 1:100. Use 10ul of the suggested working dilution to label 10^6 cells or 100ul whole blood Method sheets are available on request. For IHC-Fr: Use at a dilution of 1:200 - 1:2,000. Not recommended for the use of IHC-P. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR