Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD36 Mouse Monoclonal Antibody [clone: TR9]

Supplier: Genetex

GTX78402
89366-626EA 268.89 USD InStock
89366-626
Anti-CD36 Mouse Monoclonal Antibody [clone: TR9]
Antibodies
Monoclonal Antibody to CD36 Purified Antibody

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: TR9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty