Anti-CD36 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX112891
89357-218EA 427.1 USD
89357-218
Anti-CD36 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD36 (CD36 molecule (thrombospondin receptor))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD36
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR