Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND-10]

Supplier: GeneTex
GTX73823
89353-286EA 967.9 USD
89353-286
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND-10]
Antibodies
Monoclonal Mouse Antibody to Human Endothelial Cell CD34

For IHC-P: Use at 1:25-1:100 in an ABC method. For WB: Use at 1:100-1:500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR