Anti-CD34 Mouse Monoclonal Antibody [clone: BI-3C5]

GTX26330
89361-904EA 376.78 USD
89361-904
Anti-CD34 Mouse Monoclonal Antibody [clone: BI-3C5]
Mouse monoclonal [BI-3C5] to CD34

For WB: Tested against recombinant protein only. IHC-Fr: Use at a dilution of 1:400. Not tested in other applications. Expected to work on FACS at a dilution of 1:400 (10?l per million cells) but not yet tested in house. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: BI-3C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR