Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4]

Supplier: GeneTex
GTX78360
89366-612EA 517.02 USD
89366-612
Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4]
Antibodies
Monoclonal Antibody to CD33 (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry. Suggested working dilution is 1:1000. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: HIM3-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR