Anti-CD31 Mouse Monoclonal Antibody [clone: MEM-05]

Supplier: GeneTex
GTX22210
89361-178EA 632.51 USD
89361-178
Anti-CD31 Mouse Monoclonal Antibody [clone: MEM-05]
Antibodies
CD31 antibodies are a superior marker for angiogenesis, when compared to Factor VIII and CD34 antibodies. CD31 has been used to measure angiogenesis in association with tumor recurrence. CD31 used in a panel with CD34 and Factor VIII has also been used to mark Kaposi's sarcoma and angiosarcomas. Other studies have also indicated that CD31 and CD34 can be used as markers for myeloid progenitor cells and recognize different subsets of myeloid leukemia infiltrates (granular sarcomas).

FACS: Use at an assay dependant concentration. WB: Use at an assay dependant dilution. Use under non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: MEM-05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR