Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD31 Mouse Monoclonal Antibody [clone: B493 (158-2B3)]

Supplier: Genetex
GTX72197
89352-966
Anti-CD31 Mouse Monoclonal Antibody [clone: B493 (158-2B3)]
Mouse monoclonal [B493] to PECAM1

For FACS: Use at 1:150-1:300. For IP: Use at an assay dependent dilution. For IHC-Fr: Use at 1:50-1:100. For IHC-P: Use at 1:50-1:100. When using with formalin-fixed paraffin-embedded tissue sections an antigen demasking treatment is recommended. The optimal conditions should be determined by the individual laboratory.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: B493 (158-2B3)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty