Anti-TNFRSF8 Goat Polyclonal Antibody

Supplier: GeneTex

GTX10506
89354-084EA 708.81 USD
89354-084
Anti-TNFRSF8 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to CD30

ELISA: Use at a concentration of 0.5 - 1 ?g/ml. The detection limit for recombinant mouse CD30 ligand is approximately 0.5 ng/well. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. The detection limit for recombinant mouse CD30 ligand is 5 ng/lane under non-reducing and reducing conditions. Recombinant mature mouse CD30L has a calculated molecular mass of approximately 22 kDa. Due to glycosylation, the recombinant protein migrates to approximately 30- 45 kDa in SDS-PAGE. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNFRSF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR