Anti-CD3 Mouse Monoclonal Antibody [clone: MEM-92]

Supplier: Genetex

GTX23985
89352-072EA 395.37 USD
89352-072
Anti-CD3 Mouse Monoclonal Antibody [clone: MEM-92]
Antibodies
Mouse monoclonal [MEM - 92] to CD3

FACS: Use at an assay dependent concentration. IP: Use at an assay dependent concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: MEM-92
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR