Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3 Mouse Monoclonal Antibody

Supplier: Genetex
GTX78246
89353-824
Anti-CD3 Mouse Monoclonal Antibody
Monoclonal Antibody to CD3 (Human) Phycoerythrin (PE) conjugated

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 _l reagent / 100 _l of whole blood or 106 cells in a suspension. The content of a vial (2 ml) is sufficient for 100 tests.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty