Anti-DPP4 Chicken Polyclonal Antibody

Supplier: GeneTex
GTX17538
89367-490EA 693.18 USD
89367-490
Anti-DPP4 Chicken Polyclonal Antibody
Antibodies
CD26 [also known as dipeptidyl peptidase IV (DPP IV), adenosine deaminase (ADA) binding protein] is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross linking of CD26 using this antibody dramatically enhances the anti CD3 induced IL2 production.

For ICC: Use at a dilution of 1:100 - 1:300. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DPP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR