Anti-CD24 Mouse Monoclonal Antibody [clone: SN3]

Supplier: GeneTex
GTX74948
89353-336EA 501.7 USD
89353-336
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3]
Antibodies
Mouse monoclonal [SN3] to CD24 (RPE)

Flow Cytometry:Use 20ul of the suggested working dilution to label 10^6 cells or 100ul whole blood. Not tested in applications other than those listed above. Optimal dilutions / concentrations should be determined by the end user.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR