Anti-CD235A Mouse Monoclonal Antibody [clone: HIR2]

Supplier: GeneTex
GTX78381
89353-842EA 529.76 USD
89353-842
Anti-CD235A Mouse Monoclonal Antibody [clone: HIR2]
Antibodies
Monoclonal Antibody to CD234a Phycoerythrin (PE)

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension. The content of a vial (2 ml) is sufficient for 100 tests.

Type: Primary
Antigen: CD235A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR