Anti-CD22 Mouse Monoclonal Antibody [clone: MEM-01]

Supplier: Genetex

GTX22526
89361-268EA 395.29 USD
89361-268
Anti-CD22 Mouse Monoclonal Antibody [clone: MEM-01]
Antibodies
CD22 (BL-CAM) is a type 1 integral membrane glycoprotein with molecular weight of 130 to 140kD. It is expressed in both the cytoplasm and cell membrane of B-lymphocytes. CD22 antigen appears early in B-cell lymphocyte differentiation at approximately the same stage as the CD19 antigen. Unlike other B-cell markers, CD22 membrane expression is limited to the late differentiation stages comprised between mature B cells (CD22+) and plasma cells (CD22-), and may thus prove useful in phenotyping mature leukemias. CD22 is also strongly expressed in hairy cell leukemia.

FACS: Use at an assay dependant dilution. IP: Use at an assay dependant dilution. WB: Use at an assay dependant dilution. Detects a band of approximately 140 kDa. Use under non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: MEM-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR