Anti-CD22 Mouse Monoclonal Antibody [clone: FPC1]

GTX75381
89365-782EA 237.58 USD
89365-782
Anti-CD22 Mouse Monoclonal Antibody [clone: FPC1]
Antibodies
Mouse monoclonal antibody [FPC1] to CD22

Recommended Starting Dilutions: Immunohistology - Paraffin: 1/20 - 1/40 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: FPC1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR