Anti-CD200 Rat Monoclonal Antibody [clone: MRC OX90]

Supplier: GeneTex
GTX75637
89365-850EA 647.26 USD
89365-850
Anti-CD200 Rat Monoclonal Antibody [clone: MRC OX90]
Antibodies
Rat monoclonal [OX90] to Mouse CD200

Flow cytometry, immunohistology, ELISA; Flow Cytometry:Use 10ul of the suggested working dilution to label 10^6 cells in 100ul.

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR