Anti-CD200 Mouse Monoclonal Antibody [clone: OX-102]

Supplier: GeneTex
GTX75640
89365-852EA 511.54 USD
89365-852
Anti-CD200 Mouse Monoclonal Antibody [clone: OX-102]
Antibodies
Mouse monoclonal antibody [OX-102] to CD200R

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR